Sodium glycocholate ≥95%

Supplier: Ambeed
Warning

A196177-250MG A196177-10G
103816-824EA 23 USD
103816-824 103847-438 77667-640 77879-239
Sodium glycocholate ≥95%
Sodium glycocholate

Precaución: Inert atmosphere, 2…8 °C.

Formula: C₂₆H₄₂NNaO₆
Boiling Pt: 692 °C (760 mm Hg)
Melting Pt: 258…261 °C
Flash Pt: 372.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00036741
CAS Number: 863-57-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR