2,5-Dihydroxybenzoic acid 98%

Supplier: Ambeed
Warning

A134962-100G A134962-100MG A134962-500G A134962-25G A134962-5G
77137-046EA 36.04 USD
77137-046 77137-048 77137-050 77298-746 77298-748
2,5-Dihydroxybenzoic acid 98%
2,5-Dihydroxybenzoic acid

Precaución: for Research Use Only

Formula: C₇H₆O₄
Boiling Pt: 406.9 °C (760 mm Hg)
Melting Pt: 204…208 °C
Flash Pt: 214 °C
Storage Temperature: Ambient
MDL Number: MFCD00002460
CAS Number: 490-79-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR