N-(3-Chloro-4-fluorophenyl)-7-methoxy-6-(3-morpholinopropoxy)quinazolin-4-amine 98%

Supplier: Ambeed
Danger

Synonyms: Gefitinib

A109874-100G A109874-10G A109874-250MG A109874-25G A109874-1G A109874-5G
103797-584EA 18 USD
103797-584 103823-390 76796-746 76796-748 76796-750 76796-752 77310-196 77310-198
N-(3-Chloro-4-fluorophenyl)-7-methoxy-6-(3-morpholinopropoxy)quinazolin-4-amine 98%
N-(3-Chloro-4-fluorophenyl)-7-methoxy-6-(3-morpholinopropoxy)quinazolin-4-amine

Precaución: for Research Use Only

Formula: C₂₂H₂₄ClFN₄O₃
MW: 446.91 g/mol
Boiling Pt: 586.8 °C (760 mm Hg)
Melting Pt: 119…120 °C
Storage Temperature: Ambient
MDL Number: MFCD04307832
CAS Number: 184475-35-2
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR