Gatifloxacin 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-Cyclopropyl-6-fluoro-8-methoxy-7-(3-methylpiperazin-1-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid
A142500-10G
A142500-50MG
A142500-1G
A142500-25G
A142500-5G
103803-170EA
27
USD
103803-170
76996-912
76996-914
77316-394
77316-396
77316-398
Gatifloxacin 98%
Gatifloxacin
Precaución: for Research Use Only
Formula:
C₁₉H₂₂FN₃O₄ MW: 375.40 g/mol Boiling Pt: 607.8±55.0 °C (760 mm Hg) Melting Pt: 167…168 °C Flash Pt: 321.4 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00895399 CAS Number: 112811-59-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...