Furan-2,5-diyldimethanol 98%

Supplier: Ambeed
Warning

A136022-10G A136022-250MG A136022-500G A136022-1G A136022-25G A136022-5G A136022-100G
103802-262EA 16 USD
103802-262 76767-504 76767-506 76767-508 77315-142 77315-144 77315-146 77596-876
Furan-2,5-diyldimethanol 98%
Furan-2,5-diyldimethanol

Precaución: for Research Use Only

Formula: C₆H₈O₃
Boiling Pt: 275.4 °C (760 mm Hg)
Melting Pt: 74…77 °C
Flash Pt: 120 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003253
CAS Number: 1883-75-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR