Furan-2,5-dicarboxylic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2,5-Furandicarboxylic acid
A157618-100G
A157618-10G
A157618-1MG
A157618-500G
A157618-1G
A157618-25G
A157618-5G
A157618-1000G
103805-026EA
6
USD
103805-026
103843-228
77096-060
77096-062
77096-064
77096-066
77300-276
77319-064
77319-066
77549-990
Furan-2,5-dicarboxylic acid 98%
Furan-2,5-dicarboxylic acid
Precaución: for Research Use Only
Formula:
C₆H₄O₅ MW: 156.09 g/mol Boiling Pt: 419.2 °C (760 mm Hg) Flash Pt: 75.55 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016582 CAS Number: 3238-40-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...