2,5-Diformylfuran 98%

Supplier: Ambeed
Warning

Synonyms: Furan-2,5-dicarbaldehyde, 2,5-Furandicarboxaldehyde

A145215-100MG A145215-10G A145215-250MG A145215-5G A145215-1G A145215-25G
103803-508EA 15 USD
103803-508 103841-590 77095-588 77095-590 77095-592 77299-438 77316-874 77566-928
2,5-Diformylfuran 98%
2,5-Diformylfuran

Precaución: For research use only.

Formula: C₆H₄O₃
MW: 124.10 g/mol
Melting Pt: 108…110 °C
Flash Pt: 124.1 °C
Storage Temperature: Freezer
MDL Number: MFCD00671517
CAS Number: 823-82-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR