2(5H)-Furanone 97%

Supplier: Ambeed
Warning

Synonyms: Furan-2(5H)-one

A134936-1G A134936-25G A134936-5G A134936-100G A134936-500G
103802-074EA 6 USD
103802-074 77298-744 77314-842 77314-844 77549-620 77615-080
2(5H)-Furanone 97%
2(5H)-Furanone

Precaución: for Research Use Only

Formula: C₄H₄O₂
MW: 84.07 g/mol
Boiling Pt: 203.7 °C (760 mm Hg)
Melting Pt: 4…5 °C
Flash Pt: 87.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005376
CAS Number: 497-23-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR