(R)-2-Amino-3-((3-hydroxypropyl)thio)propanoic acid 98%

Supplier: Ambeed
Warning

A166671-100G A166671-100MG A166671-50MG A166671-1G
103806-088EA 8 USD
103806-088 103844-352 76799-232 76799-234 76799-236 77320-628
(R)-2-Amino-3-((3-hydroxypropyl)thio)propanoic acid 98%
(R)-2-Amino-3-((3-hydroxypropyl)thio)propanoic acid

Precaución: for Research Use Only

Formula: C₆H₁₃NO₃S
MW: 179.23 g/mol
Boiling Pt: 354.5 °C (760 mm Hg)
Melting Pt: 200…202 °C
Storage Temperature: Freezer
MDL Number: MFCD00899873
CAS Number: 13189-98-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR