Formamidine hydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: Formimidamide hydrochloride

A276520-100G A276520-10G A276520-500G A276520-1G A276520-25G A276520-5G
103813-308EA 9 USD
103813-308 103853-242 76802-114 76802-116 76802-118 77093-102 77093-104 77093-106
Formamidine hydrochloride 97%
Formamidine hydrochloride

Precaución: for Research Use Only

Formula: CH₄N₂
MW: 80.52 g/mol
Melting Pt: 84…87 °C
Storage Temperature: Ambient
MDL Number: MFCD00012865
CAS Number: 6313-33-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR