Formamidine acetate 95%
Supplier: Ambeed
|
Warning
|
A177386-1000G
A177386-100G
A177386-500G
103809-104EA
57
USD
103809-104
77082-948
77082-950
77111-268
77111-270
Formamidine acetate 95%
Formamidine acetate
Formula:
C₃H₈N₂O₂ Melting Pt: 158…161 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012866 CAS Number: 3473-63-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...