Fmoc-6-Ahx-OH 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Fmoc-ε-aminocaproic acid, Fmoc-ε-Acp-OH, 6-(Fmoc-amino)hexanoic acid
A162112-100G
A162112-10G
A162112-1G
A162112-500G
A162112-25G
A162112-5G
103843-826EA
9
USD
103843-826
76872-062
76872-064
76872-066
76872-068
77319-858
77319-860
Fmoc-6-Ahx-OH 97%
Fmoc-6-Ahx-OH
Precaución: for Research Use Only
Formula:
C₂₁H₂₃NO₄ MW: 353.418 g/mol Melting Pt: 114…118 °C Flash Pt: 306.2 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00077045 CAS Number: 88574-06-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...