Fmoc-L-β-homoproline 98%

Supplier: Ambeed
Warning

Synonyms: Fmoc-β-HomoPro-OH, Fmoc-β-HoPro-OH

A113093-100MG A113093-250MG A113093-1G A113093-10G
103798-162EA 36 USD
103798-162 103824-002 76995-338 76995-340 77310-838 77880-911
Fmoc-L-β-homoproline 98%
Fmoc-L-beta-homoproline

Precaución: for Research Use Only

Formula: C₂₁H₂₁NO₄
Boiling Pt: 562.1 °C (760 mm Hg)
Flash Pt: 293.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01863058
CAS Number: 193693-60-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR