Fmoc-L-β-homomethionine 98%

Supplier: Ambeed
Warning

A315738-1G A315738-5G A315738-100MG A315738-250MG
103825-082EA 60 USD
103825-082 103836-004 76838-834 76838-836 77002-770 77002-772
Fmoc-L-β-homomethionine 98%
Fmoc-L-beta-homomethionine

Precaución: for Research Use Only

Formula: C₂₁H₂₃NO₄S
Boiling Pt: 616.7 °C (760 mm Hg)
Flash Pt: 326.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01862844
CAS Number: 266359-48-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR