Fmoc-L-β-homoalanine 97%

Supplier: Ambeed
Warning

Synonyms: Fmoc-β-HoAla-OH, Fmoc-β-HomoAla-OH

A243664-100MG A243664-250MG A243664-1G A243664-5G
103811-534EA 24 USD
103811-534 103851-356 77000-502 77000-504 77090-560 77090-562
Fmoc-L-β-homoalanine 97%
Fmoc-L-beta-homoalanine

Precaución: for Research Use Only

Formula: C₁₉H₁₉NO₄
MW: 325.37 g/mol
Boiling Pt: 555.3 °C (760 mm Hg)
Flash Pt: 289.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00270346
CAS Number: 193954-26-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR