Fmoc-val-OSu 95%

Supplier: Ambeed
Warning

Synonyms: N-Fmoc-L-valine N-succinimidyl ester, Fmoc-Val-OSu

A112178-100G A112178-10G A112178-1G A112178-250MG A112178-25G A112178-500G A112178-5G
103823-808EA 10 USD
103823-808 76796-922 76796-924 76796-926 76796-928 76796-930 76796-932 77284-218
Fmoc-val-OSu 95%
Fmoc-val-OSu

Precaución: for Research Use Only

Formula: C₂₄H₂₄N₂O₆
Storage Temperature: Refrigerator
MDL Number: MFCD00153370
CAS Number: 130878-68-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR