N-Fmoc-L-valine 97%

Supplier: Ambeed
Warning

A205270-25G A205270-100G A205270-10G A205270-500G A205270-5G A205270-1000G
103848-288EA 5 USD
103848-288 77086-496 77274-984 77274-986 77274-988 77302-952 77567-970
N-Fmoc-L-valine 97%
N-Fmoc-L-valine

Precaución: For research use only.

Formula: C₂₀H₂₁NO₄
MW: 339.4 g/mol
Boiling Pt: 551.8 °C (760 mm Hg)
Melting Pt: 143…145 °C
Flash Pt: 287.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00037124
CAS Number: 68858-20-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR