N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-tyrosine 98%

Supplier: Ambeed
Warning

Synonyms: Fmoc-Tyr-OH, N-Fmoc-L-tyrosine

A149629-100G A149629-10G A149629-1G A149629-25G A149629-5G
103842-186EA 6 USD
103842-186 77028-058 77028-060 77028-062 77299-742 77317-688
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-tyrosine 98%
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-tyrosine

Precaución: for Research Use Only

Formula: C₂₄H₂₁NO₅
MW: 403.43 g/mol
Boiling Pt: 672.6 °C (760 mm Hg)
Melting Pt: 182…187 °C
Flash Pt: 360.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00134890
CAS Number: 92954-90-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR