FMOC-O-tert-butyl-L-tyrosine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
N-α-Fmoc-o-t-butyl-L-Tyrosine, N-Fmoc-O-tert-butyl-L-tyrosine, Nalpha-Fmoc-O-tert-butyl-L-tyrosine, O-tert-Butyl-Nalpha-Fmoc-L-tyrosine, Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-O-tert-butyl-L-tyrosine, Fmoc-Tyr(tBu)-OH
A290914-100G
A290914-500G
A290914-25G
A290914-5G
A290914-1000G
A290914-10G
103835-024EA
5
USD
103835-024
77277-166
77277-168
77305-586
77305-588
77568-596
77571-796
FMOC-O-tert-butyl-L-tyrosine 98%
FMOC-O-tert-butyl-L-tyrosine
Precaución: For research use only.
Formula:
C₂₈H₂₉NO₅ Boiling Pt: 658.2 °C (760 mm Hg) Melting Pt: 153…156 °C Flash Pt: 351.9 °C Storage Temperature: Ambient |
MDL Number:
MFCD00037129 CAS Number: 71989-38-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...