N(α)-Fmoc-L-tryptophan 98%

Supplier: Ambeed
Warning

Synonyms: Fmoc-Trp-OH

A203213-25G A203213-5G A203213-100G A203213-10G A203213-500G
103809-386EA 455 USD
103809-386 103848-102 77086-228 77086-230 77274-926 77274-928 77274-930
N(α)-Fmoc-L-tryptophan 98%
N(α)-Fmoc-L-tryptophan

Precaución: for Research Use Only

Formula: C₂₆H₂₂N₂O₄
MW: 426.5 g/mol
Boiling Pt: 711.9 °C (760 mm Hg)
Melting Pt: 183…185 °C
Flash Pt: 384.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00037126
CAS Number: 35737-15-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR