Fmoc-Thr-OH 95%

Supplier: Ambeed
Warning

Synonyms: (2S,3R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-hydroxybutanoic acid, Fmoc-L-Threonine

A268866-10G A268866-25G A268866-5G A268866-100G A268866-500G
103852-842EA 7 USD
103852-842 76772-110 77305-022 77305-024 77550-678 77626-402
Fmoc-Thr-OH 95%
Fmoc-Thr-OH

Precaución: for Research Use Only

Formula: C₁₉H₁₉NO₅
MW: 341.358 g/mol
Boiling Pt: 596.5 °C (760 mm Hg)
Melting Pt: 115 °C
Flash Pt: 314.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00077072
CAS Number: 73731-37-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR