Fmoc-O-(benzylphospho)-L-threonine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
N-Fmoc-O-benzylphospho-L-threonine, (2S,3R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(((benzyloxy)(hydroxy)phosphoryl)oxy)butanoic acid, Fmoc-Thr(PO(OBzl)OH)-OH, Fmoc-Thr(HPO3Bzl)-OH
A197511-100MG
A197511-250MG
A197511-25G
103816-928EA
87
USD
103816-928
103847-548
76999-210
76999-212
76999-214
Fmoc-O-(benzylphospho)-L-threonine 97%
Fmoc-O-(benzylphospho)-L-threonine
Precaución: for Research Use Only
Formula:
C₂₆H₂₆NO₈P MW: 511.467 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00797870 CAS Number: 175291-56-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...