N-Fmoc-L-serine 98% (contains < 6% H₂O)

Supplier: Ambeed
Warning

Synonyms: Fmoc-Ser-OH

A114508-100G A114508-10G A114508-1G A114508-25G A114508-5G A114508-500G
103824-256EA 6 USD
103824-256 77025-996 77025-998 77026-000 77026-002 77311-068 77670-604
N-Fmoc-L-serine 98% (contains < 6% H₂O)
N-Fmoc-L-serine

Precaución: for Research Use Only

Formula: C₁₈H₁₇NO₅
MW: 327.34 g/mol
Melting Pt: 80…84 °C
Flash Pt: 316.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00051928
CAS Number: 73724-45-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR