Fmoc-Ser-Obzl 98%

Supplier: Ambeed
Warning

A199637-100G A199637-10G A199637-250MG A199637-1G A199637-5G
103817-118EA 11 USD
103817-118 103847-744 76825-510 76825-512 76825-514 77085-786 77085-788
Fmoc-Ser-Obzl 98%
Fmoc-Ser-Obzl

Precaución: for Research Use Only

Formula: C₂₅H₂₃NO₅
Boiling Pt: 656.2 °C (760 mm Hg)
Melting Pt: 98…100 °C
Flash Pt: 350.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09752867
CAS Number: 73724-46-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR