Fmoc-O-trityl-L-serine 97%

Supplier: Ambeed
Warning

Synonyms: Fmoc-Ser(Trt)-OH

A196041-100G A196041-10G A196041-5G
103847-418EA 11 USD
103847-418 76999-134 76999-136 77085-374
Fmoc-O-trityl-L-serine 97%
Fmoc-O-trityl-L-serine

Precaución: for Research Use Only

Formula: C₃₇H₃₁NO₅
Boiling Pt: 750.4 °C (760 mm Hg)
Melting Pt: 200…210 °C
Flash Pt: 407.627 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00153364
CAS Number: 111061-56-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR