Fmoc-O-(benzylphospho)-L-serine 97%

Supplier: Ambeed
Warning

A233334-250MG A233334-1G A233334-5G A233334-25G A233334-100G
103810-884EA 38 USD
103810-884 103850-644 77000-256 77089-600 77089-602 77303-960 77881-603
Fmoc-O-(benzylphospho)-L-serine 97%
Fmoc-O-(benzylphospho)-L-serine

Precaución: for Research Use Only

Formula: C₂₅H₂₄NO₈P
Melting Pt: 113…117 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00797869
CAS Number: 158171-14-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR