Fmoc-Pro-OH, (max. 7% H₂O) 98% (contains < 7% H₂O)
Supplier: Ambeed
|
Warning
|
Synonyms:
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-proline, N-Fmoc-L-proline
A116916-100G
A116916-10G
A116916-25G
A116916-500G
A116916-5G
A116916-1000G
103824-674EA
6
USD
103824-674
77271-266
77271-268
77271-270
77271-272
77297-500
77571-022
Fmoc-Pro-OH, (max. 7% H₂O) 98% (contains < 7% H₂O)
Fmoc-Pro-OH
Precaución: for Research Use Only
Formula:
C₂₀H₁₉NO₄ MW: 337.38 g/mol Boiling Pt: 548.6 °C (760 mm Hg) Melting Pt: 117…118 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00037122 CAS Number: 71989-31-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...