Fmoc-Prolinol 97%

Supplier: Ambeed
Warning

A103246-250MG A103246-1G A103246-10G
103796-512EA 20 USD
103796-512 103822-216 76994-644 77309-098 77880-838
Fmoc-Prolinol 97%
Fmoc-Prolinol

Precaución: for Research Use Only

Formula: C₂₀H₂₁NO₃
Boiling Pt: 504.6 °C (760 mm Hg)
Melting Pt: 90 °C
Flash Pt: 259 °C
Storage Temperature: Freezer
MDL Number: MFCD00235942
CAS Number: 148625-77-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR