Fmoc-L-propargylglycine 98%

Supplier: Ambeed
Warning

Synonyms: Fmoc-Pra-OH, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-propargylglycine, N-Fmoc-L-propargylglycine, (S)-2-(Fmoc-amino)-4-pentynoic acid, Fmoc-propargyl-Gly-OH

A273755-100G A273755-1G A273755-25G A273755-5G A273755-10G A273755-250MG
103813-158EA 17 USD
103813-158 103853-082 77001-754 77092-888 77092-890 77092-892 77550-696 77881-746
Fmoc-L-propargylglycine 98%
Fmoc-L-propargylglycine

Precaución: for Research Use Only

Formula: C₂₀H₁₇NO₄
Boiling Pt: 576.7 °C (760 mm Hg)
Melting Pt: 175 °C
Flash Pt: 302.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01075095
CAS Number: 198561-07-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR