Fmoc-L-phenylalanine 98%

Supplier: Ambeed
Warning

Synonyms: N-(9-Fluorenylmethoxycarbonyl)-L-phenylalanine, N-α-Fmoc-L-Phenylalanine, Fmoc-Phe-OH, N-Fmoc-L-phenylalanine

A253740-25G A253740-100G A253740-10G A253740-500G A253740-1000G
103851-962EA 9 USD
103851-962 77091-394 77276-372 77276-374 77276-376 77550-614
Fmoc-L-phenylalanine 98%
Fmoc-L-phenylalanine

Precaución: for Research Use Only

Formula: C₂₄H₂₁NO₄
MW: 387.4 g/mol
Boiling Pt: 620.1 °C (760 mm Hg)
Melting Pt: 180…187 °C
Flash Pt: 328.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00037128
CAS Number: 35661-40-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR