N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-4-nitro-L-phenylalanine 98%

Supplier: Ambeed
Warning

A112927-100G A112927-10G A112927-250MG A112927-25G A112927-1G A112927-5G
103798-132EA 7 USD
103798-132 103823-966 76869-820 76869-822 76869-824 76869-826 77310-798 77310-800
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-4-nitro-L-phenylalanine 98%
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-4-nitro-L-phenylalanine

Precaución: for Research Use Only

Formula: C₂₄H₂₀N₂O₆
Boiling Pt: 692.3 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD00057810
CAS Number: 95753-55-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR