Fmoc-4-amino-L-phenylalanine 98%

Supplier: Ambeed
Warning

Synonyms: Fmoc-Phe(4-NH₂)-OH, 4-Amino-N-Fmoc-L-phenylalanine, Fmoc-Phe(4-NH2)-OH, (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(4-aminophenyl)propanoic acid, Fmoc-4-amino-Phe-OH

A359834-1G A359834-5G A359834-100G A359834-10G A359834-25G
103826-238EA 14 USD
103826-238 103837-246 76840-436 76840-438 76876-378 76876-380 77306-704
Fmoc-4-amino-L-phenylalanine 98%
Fmoc-4-amino-L-phenylalanine

Precaución: for Research Use Only

Formula: C₂₄H₂₂N₂O₄
MW: 402.45 g/mol
Boiling Pt: 671.5 °C (760 mm Hg)
Melting Pt: 190…205 °C
Flash Pt: 359.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00237664
CAS Number: 95753-56-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR