Fmoc-4-iodo-L-phenylalanine 97%

Supplier: Ambeed
Warning

Synonyms: Fmoc-p-iodo-Phe-OH, Fmoc-Phe(4-I)-OH

A210938-10G A210938-250MG A210938-1G A210938-25G A210938-5G A210938-100G
103818-056EA 18 USD
103818-056 103848-748 76873-674 76873-676 77087-136 77303-160 77303-162 77612-076
Fmoc-4-iodo-L-phenylalanine 97%
Fmoc-4-iodo-L-phenylalanine

Precaución: for Research Use Only

Formula: C₂₄H₂₀INO₄
MW: 513.331 g/mol
Boiling Pt: 658.8 °C (760 mm Hg)
Melting Pt: 213…217 °C
Flash Pt: 352.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00672340
CAS Number: 82565-68-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR