Fmoc-4-fluoro-L-phenylalanine 95%

Supplier: Ambeed
Warning

Synonyms: N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-4-fluoro-L-phenylalanine, Fmoc-p-fluoro-Phe-OH, Fmoc-Phe(4-F)-OH, (S)-2-[[[(9H-Fluoren-9-yl)methoxy]carbonyl]amino]-3-(4-fluorophenyl)propanoic acid, N-Fmoc-4-fluoro-L-phenylalanine

A101924-100G A101924-10G A101924-250MG A101924-25G A101924-1G A101924-5G
103796-204EA 6 USD
103796-204 103821-902 76994-546 76994-548 76994-550 76994-552 77308-788 77308-790
Fmoc-4-fluoro-L-phenylalanine 95%
Fmoc-4-fluoro-L-phenylalanine

Precaución: for Research Use Only

Formula: C₂₄H₂₀FNO₄
MW: 405.425 g/mol
Boiling Pt: 623.9 °C (760 mm Hg)
Melting Pt: 188 °C
Flash Pt: 331.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00191197
CAS Number: 169243-86-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR