Fmoc-Phe(4-CN)-OH 98%

Supplier: Ambeed
Warning

Synonyms: Fmoc-4-cyano-Phe-OH

A281767-1G A281767-25G A281767-250MG
103813-570EA 18 USD
103813-570 103853-522 76837-220 76837-222 77032-178 77032-180
Fmoc-Phe(4-CN)-OH 98%
Fmoc-Phe(4-CN)-OH

Precaución: for Research Use Only

Formula: C₂₅H₂₀N₂O₄
MW: 412.445 g/mol
Melting Pt: 190…192 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00672559
CAS Number: 173963-93-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR