Fmoc-4-chloro-L-phenylalanine 97%

Supplier: Ambeed
Warning

Synonyms: Fmoc-4-chloro-Phe-OH, N-Fmoc-4-chloro-L-phenylalanine, Fmoc-Phe(4-Cl)-OH

A623430-1G A623430-100G A623430-5G A623430-10G
103840-230EA 22 USD
103840-230 76844-400 77035-280 77307-984 77573-022
Fmoc-4-chloro-L-phenylalanine 97%
Fmoc-4-chloro-L-phenylalanine

Precaución: for Research Use Only

Formula: C₂₄H₂₀ClNO₄
MW: 421.877 g/mol
Boiling Pt: 640.9 °C (760 mm Hg)
Melting Pt: 138 °C (760 mm Hg)
Flash Pt: 341.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00080281
CAS Number: 175453-08-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR