N-Fmoc-4-(trifluoromethyl)-L-phenylalanine 95%

Supplier: Ambeed
Warning

Synonyms: Fmoc-Phe(4-CF3)-OH

A240887-10G A240887-250MG A240887-1G A240887-25G A240887-5G
103811-374EA 17 USD
103811-374 103851-180 77000-426 77000-428 77090-338 77090-340 77090-342
N-Fmoc-4-(trifluoromethyl)-L-phenylalanine 95%
N-Fmoc-4-(trifluoromethyl)-L-phenylalanine

Precaución: for Research Use Only

Formula: C₂₅H₂₀F₃NO₄
MW: 455.433 g/mol
Melting Pt: 127…130 °C
Flash Pt: 328.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00797581
CAS Number: 247113-86-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR