Fmoc-L-Phe(4-Br)-OH 97%
Supplier: Ambeed
|
Warning
|
A142027-100G
A142027-10G
A142027-250MG
A142027-1G
A142027-25G
A142027-5G
103803-090EA
23
USD
103803-090
77027-606
77027-608
77027-610
77316-296
77316-298
77316-300
Fmoc-L-Phe(4-Br)-OH 97%
Fmoc-L-Phe(4-Br)-OH
Precaución: for Research Use Only
Formula:
C₂₄H₂₀BrNO₄ Boiling Pt: 660.8 °C (760 mm Hg) Melting Pt: 151.5 °C Storage Temperature: Ambient |
MDL Number:
MFCD00273460 CAS Number: 198561-04-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...