Fmoc-3-iodo-L-phenylalanine 95%

Supplier: Ambeed
Warning

Synonyms: Fmoc-Phe(3-I)-OH

A135618-100MG A135618-250MG A135618-1G
103802-184EA 176 USD
103802-184 76798-034 76798-036 77298-800
Fmoc-3-iodo-L-phenylalanine 95%
Fmoc-3-iodo-L-phenylalanine

Precaución: for Research Use Only

Formula: C₂₄H₂₀INO₄
MW: 513.34 g/mol
Boiling Pt: 674.1±55.0 °C (760 mm Hg)
Flash Pt: 361.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01632252
CAS Number: 210282-31-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR