Fmoc-Phe(2-I)-OH 98%

Supplier: Ambeed
Warning

A134599-100MG A134599-250MG A134599-1G
103802-026EA 74 USD
103802-026 77095-150 77095-152 77314-790
Fmoc-Phe(2-I)-OH 98%
Fmoc-Phe(2-I)-OH

Precaución: for Research Use Only

Formula: C₂₄H₂₀INO₄
MW: 513.32 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01632253
CAS Number: 210282-32-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR