N-Fmoc-2-fluoro-L-phenylalanine 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(2-fluorophenyl)propanoic acid, Fmoc-Phe(2-F)-OH
A171719-10G
A171719-1G
A171719-25G
A171719-5G
103806-662EA
23
USD
103806-662
103844-938
76998-238
77082-108
77301-150
77301-152
N-Fmoc-2-fluoro-L-phenylalanine 95%
N-Fmoc-2-fluoro-L-phenylalanine
Precaución: for Research Use Only
Formula:
C₂₄H₂₀FNO₄ Boiling Pt: 620.3 °C (760 mm Hg) Melting Pt: 118 °C Flash Pt: 328.9 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00672552 CAS Number: 205526-26-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...