Fmoc-Orn-OH·HCl 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-aminopentanoic acid hydrochloride, Fmoc-ornithine hydrochloride
A218412-250MG
A218412-1G
103818-710EA
6
USD
103818-710
103849-584
76800-812
77088-104
Fmoc-Orn-OH·HCl 97%
Fmoc-Orn-OH·HCl
Precaución: for Research Use Only
Formula:
C₂₀H₂₂N₂O₄·HCl MW: 390.86 g/mol Boiling Pt: 619.4 °C (760 mm Hg) Flash Pt: 328.4 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00672329 CAS Number: 201046-57-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...