(S)-2,5-Bis((((9H-fluoren-9-yl)methoxy)carbonyl)amino)pentanoic acid 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Fmoc-Orn(Fmoc)-OH, Di-fmoc-L-ornithine
A101842-10G
A101842-250MG
A101842-25G
A101842-1G
A101842-5G
103796-172EA
23
USD
103796-172
103821-866
76994-540
76994-542
76994-544
77308-760
77308-762
(S)-2,5-Bis((((9H-fluoren-9-yl)methoxy)carbonyl)amino)pentanoic acid 95%
(S)-2,5-Bis((((9H-fluoren-9-yl)methoxy)carbonyl)amino)pentanoic acid
Precaución: for Research Use Only
Formula:
C₃₅H₃₂N₂O₆ Boiling Pt: 830.7±65.0 °C (760 mm Hg) Flash Pt: 456.2 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00153362 CAS Number: 201046-59-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...