Nδ-(tert-Butoxycarbonyl)-Nɑ-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-ornithine 98% (by HPLC)

Supplier: Ambeed
Warning

Synonyms: Nδ-Boc-Nɑ-Fmoc-L-ornithine, Nδ-Boc-Nα-Fmoc-L-ornithine, Fmoc-Orn(Boc)-OH

A120784-100G A120784-10G A120784-25G A120784-500G A120784-1G A120784-5G
103799-992EA 5 USD
103799-992 76995-810 76995-812 76995-814 76995-816 77312-100 77312-102
Nδ-(tert-Butoxycarbonyl)-Nɑ-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-ornithine 98% (by HPLC)
Ndelta-(tert-Butoxycarbonyl)-Nalpha-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-ornithine

Precaución: for Research Use Only

Formula: C₂₅H₃₀N₂O₆
MW: 454.52 g/mol
Boiling Pt: 679 °C (760 mm Hg)
Melting Pt: 111…115 °C
Flash Pt: 364.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00065668
CAS Number: 109425-55-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR