Fmoc-Orn(Alloc)-OH 98%

Supplier: Ambeed
Warning

Synonyms: Fmoc-Orn(Aloc)-OH, Nδ-Allyloxycarbonyl-Nα-Fmoc-L-ornithine

A129390-100G A129390-1G A129390-5G
103801-276EA 15 USD
103801-276 77149-542 77298-336 77298-338
Fmoc-Orn(Alloc)-OH 98%
Fmoc-Orn(Alloc)-OH

Precaución: for Research Use Only

Formula: C₂₄H₂₆N₂O₆
MW: 438.48 g/mol
Boiling Pt: 683.9 °C (760 mm Hg)
Flash Pt: 367.4 °C
Storage Temperature: Freezer
MDL Number: MFCD00798637
CAS Number: 147290-11-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR