Fmoc-L-norvaline 98%

Supplier: Ambeed
Warning

Synonyms: N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-norvaline, Fmoc-Nva-OH, N-Fmoc-L-norvaline

A173624-100G A173624-1G A173624-25G A173624-10G
103806-850EA 5 USD
103806-850 103845-150 76998-310 77301-290 77301-292 77671-468
Fmoc-L-norvaline 98%
Fmoc-L-norvaline

Precaución: for Research Use Only

Formula: C₂₀H₂₁NO₄
MW: 339.391 g/mol
Melting Pt: 151…153 °C
Flash Pt: 291.2 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00155631
CAS Number: 135112-28-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR