Fmoc-N-Me-Tyr(tBu)-OH 95%

Supplier: Ambeed
Warning

Synonyms: N-α-Fmoc-N-α-methyl-O-tert-butyl-L-tyrosine

A128315-100G A128315-250MG A128315-1G A128315-25G A128315-5G A128315-10G
103801-108EA 28 USD
103801-108 76797-760 76797-762 77313-414 77313-416 77313-418 77878-931
Fmoc-N-Me-Tyr(tBu)-OH 95%
Fmoc-N-Me-Tyr(tBu)-OH

Precaución: for Research Use Only

Formula: C₂₉H₃₁NO₅
MW: 473.569 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD02684471
CAS Number: 133373-24-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR