Fmoc-N-Me-Thr-OH 98%

Supplier: Ambeed
Warning

Synonyms: (2S,3R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)-3-hydroxybutanoic acid

A154318-100MG A154318-250MG A154318-1G A154318-25G A154318-5G
103804-626EA 45 USD
103804-626 103842-812 76823-924 76823-926 77318-518 77318-520 77318-522
Fmoc-N-Me-Thr-OH 98%
Fmoc-N-Me-Thr-OH

Precaución: for Research Use Only

Formula: C₂₀H₂₁NO₅
Boiling Pt: 566.4±50.0 °C (760 mm Hg)
Flash Pt: 296.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02094575
CAS Number: 252049-06-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR