Fmoc-N-Me-Ser(tBu)-OH 97% (EE96%)
Supplier: Ambeed
|
Warning
|
Synonyms:
N-Fmoc-O-tert-butyl-N-methyl-L-serine
A595856-1G
A595856-250MG
A595856-25G
A595856-5G
103828-950EA
37
USD
103828-950
103840-090
76844-240
77005-360
77005-362
77307-906
Fmoc-N-Me-Ser(tBu)-OH 97% (EE96%)
Fmoc-N-Me-Ser(tBu)-OH
Precaución: for Research Use Only
Formula:
C₂₃H₂₇NO₅ Boiling Pt: 557.5 °C (760 mm Hg) Melting Pt: 213…218 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD02094430 CAS Number: 197632-77-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...