Fmoc-N-Me-Phe-OH 98%

Supplier: Ambeed
Warning

Synonyms: N-Fmoc-N-methyl-L-phenylalanine, (S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)-3-phenylpropanoic acid, Fmoc-N-Me-Phe-OH, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-methyl-L-phenylalanine

A170953-1G A170953-5G A170953-250MG A170953-25G A170953-100G A170953-10G
103806-564EA 6 USD
103806-564 103844-838 77081-960 77081-962 77111-004 77550-100 77567-512 77567-514
Fmoc-N-Me-Phe-OH 98%
Fmoc-N-Me-Phe-OH

Precaución: For research use only.

Formula: C₂₅H₂₃NO₄
MW: 401.462 g/mol
Boiling Pt: 595.4 °C (760 mm Hg)
Melting Pt: 143 °C
Flash Pt: 313.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00151938
CAS Number: 77128-73-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR